Learn More
Medchemexpress LLC HY-P9901 1mg Medchemexpress, Ipilimumab CAS:477202-00-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99011MG
Encompass_Preferred
Medchemexpress, HY-P9901 1mg Ipilimumab CAS:477202-00-9 Ipilimumab is a fully human monoclonal IgG1κ antibody against the cytotoxic T-lymphocyte antigen-4 (CTLA-4), an immune-inhibitory molecule expressed in activated T cells and in suppressor T regulatory cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4389