Learn More
Medchemexpress LLC HY-P9901 5mg , Ipilimumab CAS:477202-00-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99015MG
Encompass_Preferred
HY-P9901 5mg Ipilimumab CAS: 477202-00-9 Ipilimumab is a fully human monoclonal IgG1κ antibody against the cytotoxic T-lymphocyte antigen-4 (CTLA-4), an immune-inhibitory molecule expressed in activated T cells and in suppressor T regulatory cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0332