Learn More
Abcam Ipilimumab ELISA Kit, 96TEST
Supplier:  Abcam AB23765396TEST
Encompass_Preferred
Ipilimumab ELISA Kit ab237653 is used to quantify Ipilimumab with a sensitivity of 1.6 pg/ml. Biosimilar ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5187