Learn More
Medchemexpress LLC Ipn60090 dihydrochloride | 2102101-72-2 | 99.7% | 605.44 g/mol | C24H29Cl2F3N8O3 | 1 ML
SDP
Catalog No. 5000244904
Encompass_Preferred
IPN60090 dihydrochloride is a potent, selective small-molecule inhibitor of glutaminase 1 (GLS1), provided as a dihydrochloride salt for preclinical research. It is offered as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid samples, with reported high purity and documented in vitro and in vivo activity supporting use in solid tumor studies.
  • Highly selective GLS1 inhibitor with reported IC50 = 31 nM.
  • Inhibits A549 cell proliferation (IC50 = 26 nM).
  • Available as 10 mM solution in DMSO (1 mL) or multiple solid sizes for experimental flexibility.
  • High purity suitable for biochemical and cell-based assays.
  • Documented pharmacokinetic and efficacy data in preclinical models.
Catalog No. 50-002-44904 Supplier Medchemexpress LLC Supplier No. HY103671A10MM 1ML
IPN60090 dihydrochloride is a potent, selective small-molecule inhibitor of glutaminase 1 (GLS1), provided as a dihydrochloride salt for preclinical research. It is offered as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid samples, with reported high purity and documented in vitro and in vivo activity supporting use in solid tumor studies.
  • Highly selective GLS1 inhibitor with reported IC50 = 31 nM.
  • Inhibits A549 cell proliferation (IC50 = 26 nM).
  • Available as 10 mM solution in DMSO (1 mL) or multiple solid sizes for experimental flexibility.
  • High purity suitable for biochemical and cell-based assays.
  • Documented pharmacokinetic and efficacy data in preclinical models.