Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM1697P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: IPO-38. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Raji, PHA-stimulated (>12 hours) human or mouse lymphocytes. Breast, colorectal carcinomas. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Spleen cells of a patient with hairy cell leukemia. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5424