Learn More
ABclonal Technology IPO4 Rabbit pAb
SDP
Catalog No. 5000105251
Encompass_Preferred
Predicted to enable nuclear import signal receptor activity and nuclear localization sequence binding activity. Involved in DNA replication-dependent chromatin assembly, DNA replication-independent chromatin assembly, and protein import into nucleus. Located in chromatin. Part of protein-containing complex.
Catalog No. 50-001-05251 Supplier ABclonal Technology Supplier No. A15600500UL
Predicted to enable nuclear import signal receptor activity and nuclear localization sequence binding activity. Involved in DNA replication-dependent chromatin assembly, DNA replication-independent chromatin assembly, and protein import into nucleus. Located in chromatin. Part of protein-containing complex.