Learn More
Aviva Systems Biology Corp IPO8 Antibody - C-terminal region (ARP65771_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65771_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IPO8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human IPO8. Alternate names: FLJ26580, RANBP8ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-288-567