Learn More
Cayman Chemical Ipragliflozin 100MG
SDP
Catalog No. 502987875
Encompass_Preferred
A SGLT2 inhibitor (IC50 = 7.4 nM in CHO cells expressing the human cotransporter); selective for SGLT2 over SGLT1, SGLT3, SGLT4, SGLT5, and SGLT6 (IC50s = 1.9, 30.4, 15.9, 0.46, and 10.4 µM, respectively); decreases plasma levels of insulin and glucose in an oral glucose tolerance test in a mouse model of diabetes induced by high-fat diet, STZ, and nicotinamide from 0.1-3 mg/kg; decreases plasma and hepatic IL-6, TNF-α, CCL2, and CRP levels in the same model at 3 mg/kg per day for 28 days
Catalog No. 50-298-7875 Supplier Cayman Chemical Supplier No. 22287100 MG
A SGLT2 inhibitor (IC50 = 7.4 nM in CHO cells expressing the human cotransporter); selective for SGLT2 over SGLT1, SGLT3, SGLT4, SGLT5, and SGLT6 (IC50s = 1.9, 30.4, 15.9, 0.46, and 10.4 µM, respectively); decreases plasma levels of insulin and glucose in an oral glucose tolerance test in a mouse model of diabetes induced by high-fat diet, STZ, and nicotinamide from 0.1-3 mg/kg; decreases plasma and hepatic IL-6, TNF-α, CCL2, and CRP levels in the same model at 3 mg/kg per day for 28 days