Learn More
Selleck Chemical LLC Ipragliflozin
SDP
Catalog No. 5000001639
Encompass_Preferred
Ipragliflozin (ASP1941) is a highly selective sodium-glucose cotransporter 2 (SGLT2) inhibitor with an IC50 value of 7 4 nM for hSGLT2 and a 254-fold selectivity versus SGLT1
Catalog No. 50-000-01639 Supplier Selleck Chemical LLC Supplier No. S86375MG
Ipragliflozin (ASP1941) is a highly selective sodium-glucose cotransporter 2 (SGLT2) inhibitor with an IC50 value of 7 4 nM for hSGLT2 and a 254-fold selectivity versus SGLT1