Learn More
AdipoGen Ipragliflozin
Supplier: AdipoGen AGCR13546M010
Chemical. CAS 761423-87-4. Formula C21H21FO5S. MW 404.5. Ipragliflozin is an orally active, highly potent sodium glucose co-transporter 2 SGLT-2 inhibitor IC50 = 7.4nM, selective over SGLT-1, 3, 4, 5 and 6. SGLT-2 is one subtype of SGLTs found almost exclusively in the proximal tubules of nephronic components in kidneys, playing a key role in the re-uptake of glucose in the proximal tubule of the kidneys. Inhibition of SGLT-2 reduces blood glucose by blocking renal glucose reabsorption and thereby increasing urinary glucose excretion UGE. Anti-diabetic and anti-obesity agent. Shown to improve glycemic control and reduce body weight, furthermore, lowering hypoglycemic risk and abdominal symptoms. SGLT-2 inhibitors are likely to improve beta-cell function and insulin sensitivity and restore glucose homeostasis. Attenuates non-alcoholic steatohepatitis NASH development.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.