Learn More
Medchemexpress LLC Ipragliflozin L-Proline | 951382-34-6 | 99.9% | 519.58 | 200 MG
SDP
Catalog No. 5000359506
Encompass_Preferred
Ipragliflozin (L-Proline) is a highly potent and selective SGLT2 inhibitor with an IC50 of 2.8 nM, showing little to no potency for SGLT1/3/4/5/6. It is intended for research use only.
  • Potently and selectively inhibits human, rat, and mouse SGLT2.
  • Exhibits stability against intestinal glucosidases.
  • Shows good pharmacokinetic properties following oral dosing.
  • Dose-dependently increases urinary glucose excretion.
  • Reduces blood glucose and plasma insulin levels.
  • Improves glucose intolerance.
Catalog No. 50-003-59506 Supplier Medchemexpress LLC Supplier No. HY14894A200MG
Ipragliflozin (L-Proline) is a highly potent and selective SGLT2 inhibitor with an IC50 of 2.8 nM, showing little to no potency for SGLT1/3/4/5/6. It is intended for research use only.
  • Potently and selectively inhibits human, rat, and mouse SGLT2.
  • Exhibits stability against intestinal glucosidases.
  • Shows good pharmacokinetic properties following oral dosing.
  • Dose-dependently increases urinary glucose excretion.
  • Reduces blood glucose and plasma insulin levels.
  • Improves glucose intolerance.