Learn More
Selleck Chemical LLC Ipragliflozin L-Proline S0994-25MG
SDP
Catalog No. 5000042621
Encompass_Preferred
Ipragliflozin L-Proline is an orally active and selective inhibitor of SGLT2 with IC50 of 7 38 nM 6 73 nM and 5 64 nM for human SGLT2 rat SGLT2 and mouse SGLT2 respectively
Catalog No. 50-000-42621 Supplier Selleck Chemical LLC Supplier No. S099425MG
Ipragliflozin L-Proline is an orally active and selective inhibitor of SGLT2 with IC50 of 7 38 nM 6 73 nM and 5 64 nM for human SGLT2 rat SGLT2 and mouse SGLT2 respectively