Learn More
Apexbio Technology LLC Ipratropium Bromide 22254-24-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096240
Encompass_Preferred
Ipratropium bromide (CAS 22254-24-6) is a synthetic quaternary ammonium compound structurally derived from atropine It functions as a muscarinic acetylcholine receptor (mAChR) antagonist primarily inhibiting M3 receptor-mediated bronchoconstriction in airway smooth muscle By blocking parasympathetic-mediated bronchial smooth muscle contraction it induces bronchodilation Ipratropium bromide is widely utilized in research to investigate cholinergic signaling pathways airway physiology and the pharmacological modulation of bronchomotor tone Its properties make it valuable for studies focused on obstructive pulmonary conditions and respiratory drug development
Catalog No. 50-000-96240 Supplier Apexbio Technology LLC Supplier No. A84515.1
Ipratropium bromide (CAS 22254-24-6) is a synthetic quaternary ammonium compound structurally derived from atropine It functions as a muscarinic acetylcholine receptor (mAChR) antagonist primarily inhibiting M3 receptor-mediated bronchoconstriction in airway smooth muscle By blocking parasympathetic-mediated bronchial smooth muscle contraction it induces bronchodilation Ipratropium bromide is widely utilized in research to investigate cholinergic signaling pathways airway physiology and the pharmacological modulation of bronchomotor tone Its properties make it valuable for studies focused on obstructive pulmonary conditions and respiratory drug development