Learn More
Selleck Chemical LLC Ipratropium Bromide S1683-50mg
SDP
Catalog No. 503111593
Encompass_Preferred
Ipratropium Bromide is an antagonist of M3 type muscarinic acetylcholine receptors used for the treatment of chronic obstructive pulmonary disease (COPD)
Catalog No. 50-311-1593 Supplier Selleck Chemical LLC Supplier No. S168350MG
Ipratropium Bromide is an antagonist of M3 type muscarinic acetylcholine receptors used for the treatment of chronic obstructive pulmonary disease (COPD)