Learn More
Apexbio Technology LLC Ipratropium Bromide 22254-24-6 50mg
SDP
Catalog No. 502240613
Encompass_Preferred
Ipratropium bromide (CAS 22254-24-6) is a synthetic quaternary ammonium compound structurally derived from atropine It functions as a muscarinic acetylcholine receptor (mAChR) antagonist primarily inhibiting M3 receptor-mediated bronchoconstriction in airway smooth muscle By blocking parasympathetic-mediated bronchial smooth muscle contraction it induces bronchodilation Ipratropium bromide is widely utilized in research to investigate cholinergic signaling pathways airway physiology and the pharmacological modulation of bronchomotor tone Its properties make it valuable for studies focused on obstructive pulmonary conditions and respiratory drug development
This item is not returnable. View return policy
Catalog No. 50-224-0613 Supplier Apexbio Technology LLC Supplier No. A845150
Ipratropium bromide (CAS 22254-24-6) is a synthetic quaternary ammonium compound structurally derived from atropine It functions as a muscarinic acetylcholine receptor (mAChR) antagonist primarily inhibiting M3 receptor-mediated bronchoconstriction in airway smooth muscle By blocking parasympathetic-mediated bronchial smooth muscle contraction it induces bronchodilation Ipratropium bromide is widely utilized in research to investigate cholinergic signaling pathways airway physiology and the pharmacological modulation of bronchomotor tone Its properties make it valuable for studies focused on obstructive pulmonary conditions and respiratory drug development