Learn More
Medchemexpress LLC Ipratropium bromide hydrate | 66985-17-9 | 99.9% | 1 ML
SDP
Catalog No. 5000411570
Encompass_Preferred
Ipratropium bromide (Sch 1000) hydrate is a muscarinic receptor antagonist, with IC50s of 2.9 nM, 2 nM, and 1.7 nM for M1, M2, and M3 receptors, respectively. It relaxes smooth muscle and can be used in the research for COPD (chronic obstructive pulmonary disease) and asthma.
  • Muscarinic receptor antagonist
  • Relaxes smooth muscle
  • Potential for research in COPD and asthma
  • IC50s: 2.9 nM for M1, 2 nM for M2, and 1.7 nM for M3 receptors
Catalog No. 50-004-11570 Supplier Medchemexpress LLC Supplier No. HYB133210MM/1ML
Ipratropium bromide (Sch 1000) hydrate is a muscarinic receptor antagonist, with IC50s of 2.9 nM, 2 nM, and 1.7 nM for M1, M2, and M3 receptors, respectively. It relaxes smooth muscle and can be used in the research for COPD (chronic obstructive pulmonary disease) and asthma.
  • Muscarinic receptor antagonist
  • Relaxes smooth muscle
  • Potential for research in COPD and asthma
  • IC50s: 2.9 nM for M1, 2 nM for M2, and 1.7 nM for M3 receptors