Learn More
Medchemexpress LLC Ipratropium-d3 bromide | 22254-24-6 | 98.0% | 1 MG
SDP
Catalog No. 5000406155
Encompass_Preferred
Ipratropium-d3 bromide is a deuterium labeled muscarinic receptor antagonist, useful in research for chronic obstructive pulmonary disease (COPD) and asthma. It can also serve as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Muscarinic receptor antagonist
  • Can be used in research for COPD and asthma
  • Serves as a tracer
  • Functions as an internal standard for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06155 Supplier Medchemexpress LLC Supplier No. HYB0241S1MG
Ipratropium-d3 bromide is a deuterium labeled muscarinic receptor antagonist, useful in research for chronic obstructive pulmonary disease (COPD) and asthma. It can also serve as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Muscarinic receptor antagonist
  • Can be used in research for COPD and asthma
  • Serves as a tracer
  • Functions as an internal standard for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles of drugs