Learn More
Medchemexpress LLC Ipratropium-d3 bromide | 22254-24-6 | 98.0% | 5 MG
SDP
Catalog No. 5000406955
Encompass_Preferred
Ipratropium-d3 bromide is the deuterium labeled Ipratropium bromide. It is a muscarinic receptor antagonist, with binding IC50 values of 2.9 nM, 2 nM, and 1.7 nM for M1, M2, and M3 receptors, respectively. It can be used in the research for COPD (chronic obstructive pulmonary disease) and asthma, and serves as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Acts as a muscarinic receptor antagonist
  • Suitable for research on COPD and asthma
  • Functions as a tracer
  • Useful as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06955 Supplier Medchemexpress LLC Supplier No. HYB0241S5MG
Ipratropium-d3 bromide is the deuterium labeled Ipratropium bromide. It is a muscarinic receptor antagonist, with binding IC50 values of 2.9 nM, 2 nM, and 1.7 nM for M1, M2, and M3 receptors, respectively. It can be used in the research for COPD (chronic obstructive pulmonary disease) and asthma, and serves as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Acts as a muscarinic receptor antagonist
  • Suitable for research on COPD and asthma
  • Functions as a tracer
  • Useful as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS