Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ipratropium-d3 bromide | 98.0% | 415.38 | C20H27D3BrNO3 | 10 MG
SDP

Catalog No. 5000298268
Encompass_Preferred

Deuterium-labeled ipratropium bromide supplied as a solid research standard for analytical applications. It is intended for use as an internal standard or tracer in mass spectrometry and quantitative assays, providing mass-differentiation for accurate measurement of ipratropium in biological samples.

  • Deuterium-labeled internal standard for mass spectrometry
  • Suitable for analytical quantification and tracer studies
  • Reported purity 98.0%
  • Molecular formula C20H27D3BrNO3 and molecular weight 415.38
  • Store sealed at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month)

Catalog No. 50-002-98268 Supplier Medchemexpress LLC Supplier No. HYB0241S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled ipratropium bromide supplied as a solid research standard for analytical applications. It is intended for use as an internal standard or tracer in mass spectrometry and quantitative assays, providing mass-differentiation for accurate measurement of ipratropium in biological samples.

  • Deuterium-labeled internal standard for mass spectrometry
  • Suitable for analytical quantification and tracer studies
  • Reported purity 98.0%
  • Molecular formula C20H27D3BrNO3 and molecular weight 415.38
  • Store sealed at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.