Learn More
Alzet iPRECIO Micro Infusion Pumps SMP-200: Implantable, Programmable, Refillable Infusion Device for Rats, for Use with Management System IMS-200, Pack of 5 Pumps
SDP
Catalog No. NC0456226
Encompass
iPRECIO SMP-200 pumps are advanced infusion devices for drug delivery in rats and larger animals. With versatile programmable functions and a refillable reservoir, these implantable infusion pumps enable full control over dosing studies, including complex dosing protocols such as multiple bolus dosing, variable/constant delivery, escalating dose administration, and more. Access the pump reservoir in situ using the percutaneously accessible septum, which allows one to change solutions or refill the pump during the study. Pumps are EtO sterilized.Set the infusion profile with the user-friendly iPRECIO management software and then download it to the pump’s memory chip. [Please note that the management software (IMS-200) is sold separately.]iPRECIO SMP-200:•For rats and larger animals•Refillable while in vivo•IR data communication•Reservoir volume: 900 µl•Flow rates: 0-30 µl/hr[For drug delivery in mice, please see the iPRECIO SMP-310R.]
Catalog No. NC0456226 Supplier Alzet Supplier No. 0006100
iPRECIO SMP-200 pumps are advanced infusion devices for drug delivery in rats and larger animals. With versatile programmable functions and a refillable reservoir, these implantable infusion pumps enable full control over dosing studies, including complex dosing protocols such as multiple bolus dosing, variable/constant delivery, escalating dose administration, and more. Access the pump reservoir in situ using the percutaneously accessible septum, which allows one to change solutions or refill the pump during the study. Pumps are EtO sterilized.Set the infusion profile with the user-friendly iPRECIO management software and then download it to the pump’s memory chip. [Please note that the management software (IMS-200) is sold separately.]iPRECIO SMP-200:•For rats and larger animals•Refillable while in vivo•IR data communication•Reservoir volume: 900 µl•Flow rates: 0-30 µl/hr[For drug delivery in mice, please see the iPRECIO SMP-310R.]