Learn More
Medchemexpress LLC HY-N0094 1g Medchemexpress, Ipriflavone CAS:35212-22-7 Purity:>98%
SDP
Catalog No. 501933388
Encompass_Preferred
Medchemexpress, HY-N0094 1g Ipriflavone CAS:35212-22-7 Ipriflavone is a synthetic isoflavone derivative used to suppress bone resorption. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3388 Supplier Medchemexpress LLC Supplier No. HYN00941G
Medchemexpress, HY-N0094 1g Ipriflavone CAS:35212-22-7 Ipriflavone is a synthetic isoflavone derivative used to suppress bone resorption. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.