Learn More
Apexbio Technology LLC Ipriflavone (Osteofix) 35212-22-7 1g
SDP
Catalog No. 5000088960
Encompass_Preferred
Ipriflavone Osteofix is a small-molecule inhibitor targeting bone resorption It is designed to modulate osteoclast-related signaling pathways thereby reducing bone resorptive activity Ipriflavone Osteofix exerts its biological activity primarily through inhibition of osteoclast-mediated bone remodeling In in vitro studies Ipriflavone Osteofix demonstrates inhibition of cellular signaling processes involved in bone mineral loss Based on these pharmacological properties Ipriflavone Osteofix holds research potential in experimental studies of osteoporosis and related bone disorders
Catalog No. 50-000-88960 Supplier Apexbio Technology LLC Supplier No. B36731000
Ipriflavone Osteofix is a small-molecule inhibitor targeting bone resorption It is designed to modulate osteoclast-related signaling pathways thereby reducing bone resorptive activity Ipriflavone Osteofix exerts its biological activity primarily through inhibition of osteoclast-mediated bone remodeling In in vitro studies Ipriflavone Osteofix demonstrates inhibition of cellular signaling processes involved in bone mineral loss Based on these pharmacological properties Ipriflavone Osteofix holds research potential in experimental studies of osteoporosis and related bone disorders