Learn More
Apexbio Technology LLC Ipriflavone (Osteofix) 35212-22-7 500mg
SDP
Supplier:  Apexbio Technology LLC B3673500
Encompass_Preferred
Ipriflavone Osteofix is a small-molecule inhibitor targeting bone resorption It is designed to modulate osteoclast-related signaling pathways thereby reducing bone resorptive activity Ipriflavone Osteofix exerts its biological activity primarily through inhibition of osteoclast-mediated bone remodeling In in vitro studies Ipriflavone Osteofix demonstrates inhibition of cellular signaling processes involved in bone mineral loss Based on these pharmacological properties Ipriflavone Osteofix holds research potential in experimental studies of osteoporosis and related bone disorders
Catalog No. 50-224-0819