Learn More
TARGETMOL CHEMICALS INC IPRINDOLE 25MG
SDP
Catalog No. 502580152
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Iprindole a tricyclic indole antidepressant is a dual weak inhibitor of noradrenaline and 5-HT uptake [1]. purity: 99%
Catalog No. 50-258-0152 Supplier TARGETMOL CHEMICALS INC Supplier No. T3701525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Iprindole a tricyclic indole antidepressant is a dual weak inhibitor of noradrenaline and 5-HT uptake [1]. purity: 99%