Learn More
Medchemexpress LLC HY-B1978 250mg Medchemexpress, Iprodione CAS:36734-19-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1978250MG
Encompass_Preferred
Medchemexpress, HY-B1978 250mg Iprodione CAS:36734-19-7 Iprodione, a dicarboximide fungicide, has a highly specific action, with a capacity to cause oxidative damage through production of free oxygen radicals (ROS). Iprodione does not appear to be species selective. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2717