Learn More
Medchemexpress LLC HY-B0886A 100mg Medchemexpress, Iproniazid CAS:54-92-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0886A100MG
Encompass_Preferred
Medchemexpress, HY-B0886A 100mg Iproniazid CAS:54-92-2 Iproniazid is a non-selective, irreversible monoamine oxidase (MAO) inhibitor of the hydrazine class. Iproniazid has antidepressive activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1139