Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iproniazid Phosphate 97%, Thermo Scientific™

Catalog No. AC122470250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC122470250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC122470250 Supplier Thermo Scientific Chemicals Supplier No. 122470250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-33-9
Molecular Formula C9H16N3O5P
Molecular Weight (g/mol) 277.217
InChI Key YPDVTKJXVHYWFY-UHFFFAOYSA-N
Synonym iproniazid phosphate, iproniazide phosphate, marsilid phosphate, isoproniazid phosphate, iproniazid as phosphate, n'-isopropylisonicotinohydrazide phosphate, iproniazid dihydrogen phosphate, iproniazid phosphate van, unii-8de00v62tv
PubChem CID 9367
IUPAC Name phosphoric acid;N'-propan-2-ylpyridine-4-carbohydrazide
SMILES CC(C)NNC(=O)C1=CC=NC=C1.OP(=O)(O)O

Specifications

Quantity 25 g
Merck Index 15, 5122
Formula Weight 277.2
Chemical Name or Material Iproniazid phosphate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.