Learn More
Cayman Chemical Ipronidazole, Purity: ≥90%, CAS: 14885-29-1, 50 mg
SDP
Catalog No. 502911432
Encompass_Preferred
A nitroimidazole antiprotozoal agent; dietary administration reduces H. meleagridis-induced mortality in young turkeys from 0.00312-0.0094% w/w; induces 100% disease clearance in bulls infected with T. foetus at 60 g/animal
Catalog No. 50-291-1432 Supplier Cayman Chemical Supplier No. 3508250 MG
A nitroimidazole antiprotozoal agent; dietary administration reduces H. meleagridis-induced mortality in young turkeys from 0.00312-0.0094% w/w; induces 100% disease clearance in bulls infected with T. foetus at 60 g/animal