Learn More
Cayman Chemical IpronIdazole-d3 500ug
SDP
Catalog No. 502915429
Encompass_Preferred
An internal standard for the quantification of ipronidazole by GC- or LC-MS
Catalog No. 50-291-5429 Supplier Cayman Chemical Supplier No. 37194500 UG
An internal standard for the quantification of ipronidazole by GC- or LC-MS