Learn More
Bio Rad Laboratories IPROOF HF MASTER MIX
Catalog No. 509012992
Encompass
Mix, Master; Bio-Rad; iProof HF; For high fidelity templates; 100 x 50uL reactions; Includes 2X master mix (0.04 U/uL), DMSO
Catalog No. 50-901-2992 Supplier Bio Rad Laboratories Supplier No. 1725310
Mix, Master; Bio-Rad; iProof HF; For high fidelity templates; 100 x 50uL reactions; Includes 2X master mix (0.04 U/uL), DMSO