Learn More
Bio Rad Laboratories IPROOF HIGHFIDELITY PCR KIT
Catalog No. 509010770
Encompass
Kit, iProof High-Fidelity PCR; Bio-Rad; Size: Size: 2 U/uL; 50 U; Includes 5x reaction buffers, MgCl2 solution, DMSO, dNTPs, lambda DNA, 1.3 and 10 kb primers, DNA standard
Catalog No. 50-901-0770 Supplier Bio Rad Laboratories Supplier No. 1725330
Kit, iProof High-Fidelity PCR; Bio-Rad; Size: Size: 2 U/uL; 50 U; Includes 5x reaction buffers, MgCl2 solution, DMSO, dNTPs, lambda DNA, 1.3 and 10 kb primers, DNA standard