Learn More
Alkali Scientific IPS2050-20 Overhead Stirrer OS
SDP
Catalog No. 503029613
Encompass_Preferred
LCD control panel Includes complete stand assembly Compact and quiet Digital accuracy and convenience The Alkali ScientificIPS2050-20 Overhead Stirrers OS20L feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples Speed and run time are accurately set and monitored on the LCD screen The 20L capacity models are supplied with a 4 arm stainless steel propeller additional steel and Teflon accessories are available A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction while a splash cover protects the connection mechanism from potential corrosion Unlike most mixers the Alkali Industrial series is also supplied with a support stand and mounting hardware The stirrer easily and securely attaches to the stand for compatibility with a variet
Catalog No. 50-302-9613 Supplier Alkali Scientific Supplier No. RS2050
LCD control panel Includes complete stand assembly Compact and quiet Digital accuracy and convenience The Alkali ScientificIPS2050-20 Overhead Stirrers OS20L feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples Speed and run time are accurately set and monitored on the LCD screen The 20L capacity models are supplied with a 4 arm stainless steel propeller additional steel and Teflon accessories are available A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction while a splash cover protects the connection mechanism from potential corrosion Unlike most mixers the Alkali Industrial series is also supplied with a support stand and mounting hardware The stirrer easily and securely attaches to the stand for compatibility with a variet