Learn More
Alkali Scientific Alkali Scientific™ IPS2050-40 Overhead Stirrer OS40L, 40L includes stand, and 4 arm propeller, 115V
SDP
Catalog No. 503029653
Encompass_Preferred
The Alkali Scientific™ IPS2050-40 Overhead Stirrers OS40L feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples.  Speed and run time are accurately set and monitored on the LCD screen.  The 40L  capacity models are supplied with a 4 arm stainless steel propeller, additional steel and Teflon accessories are available.  A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools.  The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Alkali Industrial series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.
Catalog No. 50-302-9653 Supplier Alkali Scientific Supplier No. RS20501
The Alkali Scientific™ IPS2050-40 Overhead Stirrers OS40L feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples.  Speed and run time are accurately set and monitored on the LCD screen.  The 40L  capacity models are supplied with a 4 arm stainless steel propeller, additional steel and Teflon accessories are available.  A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools.  The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Alkali Industrial series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.