Learn More
Medchemexpress LLC Ipsapirone | 95847-70-4 | 99.9% | 401.49 | 5 MG
SDP
Catalog No. 5000402718
Encompass_Preferred
Ipsapirone is an anxiolytic compound that acts as a partial agonist for the 5-HT1A receptor. It also demonstrates an antagonistic effect on the 5-HT1A receptor and, at higher doses, can inhibit 5-HT2 and α1-adrenergic functions.
  • Functions as a 5-HT1A receptor partial agonist.
  • Exhibits 5-HT1A receptor antagonistic effects.
  • Can inhibit 5-HT2 and α1-adrenergic function at high doses.
  • Available in solid form, white to off-white in color.
  • Can be stored at room temperature for shipping in the continental US.
  • Powder form is stable for 3 years at -20°C.
  • Solution form is stable for 6 months at -80°C and 1 month at -20°C.
Catalog No. 50-004-02718 Supplier Medchemexpress LLC Supplier No. HY196865MG
Ipsapirone is an anxiolytic compound that acts as a partial agonist for the 5-HT1A receptor. It also demonstrates an antagonistic effect on the 5-HT1A receptor and, at higher doses, can inhibit 5-HT2 and α1-adrenergic functions.
  • Functions as a 5-HT1A receptor partial agonist.
  • Exhibits 5-HT1A receptor antagonistic effects.
  • Can inhibit 5-HT2 and α1-adrenergic function at high doses.
  • Available in solid form, white to off-white in color.
  • Can be stored at room temperature for shipping in the continental US.
  • Powder form is stable for 3 years at -20°C.
  • Solution form is stable for 6 months at -80°C and 1 month at -20°C.