Learn More
Cayman Chemical IpsapIronhydrochlorIde 50mg
SDP
Catalog No. 502987594
Encompass_Preferred
A partial agonist of serotonin 5-HT1A receptors (Ki = 10 nM in hippocampal membranes); reduces 5-HT release in rat ventral hippocampus in vivo; low doses decrease, while high doses increase, extracellular dopamine release in murine nucleus accumbens; extracellular dopamine levels in murine striatum increase following administration at concentrations >0.1 mg/kg; has anxiolytic effects in vivo, inhibiting foot shock-induced aggression and passive avoidance behavior in rats (ED50s = 2.2 and 0.5 mg/kg, respectively)
Catalog No. 50-298-7594 Supplier Cayman Chemical Supplier No. 2207550 MG
A partial agonist of serotonin 5-HT1A receptors (Ki = 10 nM in hippocampal membranes); reduces 5-HT release in rat ventral hippocampus in vivo; low doses decrease, while high doses increase, extracellular dopamine release in murine nucleus accumbens; extracellular dopamine levels in murine striatum increase following administration at concentrations >0.1 mg/kg; has anxiolytic effects in vivo, inhibiting foot shock-induced aggression and passive avoidance behavior in rats (ED50s = 2.2 and 0.5 mg/kg, respectively)