Learn More
Medchemexpress LLC HY-127105 10mg , Iptacopan CAS:1644670-37-0 Purity:>98%
SDP
Catalog No. 502259479
Encompass_Preferred
HY-127105 10mg Iptacopan CAS: 1644670-37-0 Iptacopan (LNP023) is a first-in-class, orally bioavailable, highly potent and highly selective factor B inhibitor with an IC50 value of 10 nM. Iptacopan shows direct, reversible, and high-affinity binding to human factor B with a KD of 7.9 nM. Iptacopan targets the underlying cause of complement 3 glomerulopathy (C3G). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9479 Supplier Medchemexpress LLC Supplier No. HY12710510MG
HY-127105 10mg Iptacopan CAS: 1644670-37-0 Iptacopan (LNP023) is a first-in-class, orally bioavailable, highly potent and highly selective factor B inhibitor with an IC50 value of 10 nM. Iptacopan shows direct, reversible, and high-affinity binding to human factor B with a KD of 7.9 nM. Iptacopan targets the underlying cause of complement 3 glomerulopathy (C3G). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.