Learn More
Medchemexpress LLC HY-127105 5mg Medchemexpress, Iptacopan CAS:1644670-37-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1271055MG
Encompass_Preferred
Medchemexpress, HY-127105 5mg Iptacopan CAS:1644670-37-0 Iptacopan (LNP023) is a first-in-class, orally bioavailable, highly potent and highly selective factor B inhibitor with an IC50 value of 10 nM. Iptacopan shows direct, reversible, and high-affinity binding to human factor B with a KD of 7.9 nM. Iptacopan targets the underlying cause of complement 3 glomerulopathy (C3G). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1125