Learn More
Selleck Chemical LLC Iptacopan Hydrochloride
SDP
Supplier:  Selleck Chemical LLC E0119100MG
Encompass_Preferred
Iptacopan Hydrochloride is a first-in-class orally administered potent and highly selective factor B inhibitor with an IC50 of 12 nM and a Kd value of 7 9 nM Iptacopan exhibits excellent selectivity over other proteases affording IC50s of 30 M across a panel of 41 human proteases
Catalog No. 50-313-6008