Learn More
Biotang Inc IPTG 10 GM, P-806, CAS # 367-93-1. Purity > 99%, soluble in ethanol and water. Shipped at room temperature. Store at 2-8 degrees Celsius. Dioxane free. Mol. wt. 238.3.
Catalog No. 50152356 Shop All BIOTANG Inc Products
Encompass_Preferred
IPTG 10 GM, P-806, CAS # 367-93-1. Purity > 99%, soluble in ethanol and water. Shipped at room temperature. Store at 2-8 degrees Celsius. Dioxane free. Mol. wt. 238.3.
Catalog No. 50-152-356 Supplier Biotang Inc Supplier No. BTBB902
IPTG 10 GM, P-806, CAS # 367-93-1. Purity > 99%, soluble in ethanol and water. Shipped at room temperature. Store at 2-8 degrees Celsius. Dioxane free. Mol. wt. 238.3.