Learn More
Selleck Chemical LLC IPTG-100mg
SDP
Supplier:  Selleck Chemical LLC S6826100MG
Encompass_Preferred
IPTG (Isopropyl β-D-thiogalactoside, Isopropyl thiogalactoside, Isopropyl β-D-1-thiogalactopyranoside, lad-y) is a galactose and a molecular mimic of allolactose that triggers transcription of the lac operon, stimulating protein expression.
Catalog No. 50-238-9950