Learn More
Gold Biotechnology Inc IPTG 100 g
SDP
Catalog No. 501532802
Encompass_Preferred
IPTG is an analog of galactose that is non-metabolizable and inactivates the lac repressor to induce synthesis of β-galactosidase in E. coli. The expression of cloned genes under the control of the lac operon is induced by IPTG. It is also a substrate for thigalactoside transacetylase and has been reported to induce penicillinase in bacteria.
Catalog No. 50-153-2802 Supplier Gold Biotechnology Inc Supplier No. I2481C100
IPTG is an analog of galactose that is non-metabolizable and inactivates the lac repressor to induce synthesis of β-galactosidase in E. coli. The expression of cloned genes under the control of the lac operon is induced by IPTG. It is also a substrate for thigalactoside transacetylase and has been reported to induce penicillinase in bacteria.