Learn More
Ubpbio IPTG, 10G
Catalog No. 501126934
Encompass_Preferred
Protein Purification Reagents – IPTG (UBPBio catalog# P1010-10, P1010-25, P1010-100) – IPTG, Isopropyl β-D-1-thiogalactopyranoside, binds the lac repressor and induces expression of β-galactosidase in E. coli. IPTG is widely used to induce the expression of cloned genes under the control of the lac operon and to screen blue/white colony in X-gal plate.
Catalog No. 50-112-6934 Supplier Ubpbio Supplier No. P101010
Protein Purification Reagents – IPTG (UBPBio catalog# P1010-10, P1010-25, P1010-100) – IPTG, Isopropyl β-D-1-thiogalactopyranoside, binds the lac repressor and induces expression of β-galactosidase in E. coli. IPTG is widely used to induce the expression of cloned genes under the control of the lac operon and to screen blue/white colony in X-gal plate.