Learn More
Selleck Chemical LLC IPTG
SDP
Catalog No. 5000002628
Encompass_Preferred
IPTG (Isopropyl -D-thiogalactoside Isopropyl thiogalactoside Isopropyl -D-1-thiogalactopyranoside lad-y) is a galactose and a molecular mimic of allolactose that triggers transcription of the lac operon stimulating protein expression
Catalog No. 50-000-02628 Supplier Selleck Chemical LLC Supplier No. S68261G
IPTG (Isopropyl -D-thiogalactoside Isopropyl thiogalactoside Isopropyl -D-1-thiogalactopyranoside lad-y) is a galactose and a molecular mimic of allolactose that triggers transcription of the lac operon stimulating protein expression