Learn More
Gold Biotechnology Inc IPTG 300 g
SDP
Catalog No. 5000562095
Encompass_Preferred
IPTG is an analog of galactose that is non-metabolizable and inactivates the lac repressor to induce synthesis of -galactosidase in E coli The expression of cloned genes under the control of the lac operon is induced by IPTG It is also a substrate for thigalactoside transacetylase and has been reported to induce penicillinase in bacteria
Catalog No. 50-005-62095 Supplier Gold Biotechnology Inc Supplier No. I2481C300
IPTG is an analog of galactose that is non-metabolizable and inactivates the lac repressor to induce synthesis of -galactosidase in E coli The expression of cloned genes under the control of the lac operon is induced by IPTG It is also a substrate for thigalactoside transacetylase and has been reported to induce penicillinase in bacteria