Learn More
Research Products International Corp IPTG 300 G
SDP
Catalog No. 502230419
Encompass_Preferred
IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.
Dioxane-Free
Water Content: <0.49%
Catalog No. 50-223-0419 Supplier Research Products International Corp Supplier No. I56000300.0
IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.
Dioxane-Free
Water Content: <0.49%