Learn More
Biotium IPTG, 5 g
Catalog No. 501964098
Encompass_Preferred
IPTG (Isopropylthio-β-galactoside) is an inducer of β-galactosidase activity in bacteria and is suitable for use with X-gal or bluo-gal to detect lac gene activity in cloning procedures. Properties: White solid soluble in DMSO; Store desiccated at 4°C and protect from light; C9H18O5S; MW: 238.3
Catalog No. 50-196-4098 Supplier Biotium Supplier No. 100212
IPTG (Isopropylthio-β-galactoside) is an inducer of β-galactosidase activity in bacteria and is suitable for use with X-gal or bluo-gal to detect lac gene activity in cloning procedures. Properties: White solid soluble in DMSO; Store desiccated at 4°C and protect from light; C9H18O5S; MW: 238.3