Learn More
AG Scientific Inc IPTG, 500 G
Catalog No. 502237093
Encompass_Preferred
IPTG known as Isopropyl-Beta-D-Thiogalactopyranoside functions as an inducer of galactosidase activity by binding to and inhibiting the repressor.It is utilized for the induction of expression from the lac promoter and derivates. IPTG is also used to differentiate recombinants from non-recombinants in cloning strategies using vectors containing the Z or Z peptide gene.CAS Number: 367-93-1Molecular Weight: 238.3 DaChemical Formula: C9H18O5SSolubility: Soluble in water (5%), renders clear colorless solutionStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7093 Supplier AG Scientific Inc Supplier No. I1312500GM
IPTG known as Isopropyl-Beta-D-Thiogalactopyranoside functions as an inducer of galactosidase activity by binding to and inhibiting the repressor.It is utilized for the induction of expression from the lac promoter and derivates. IPTG is also used to differentiate recombinants from non-recombinants in cloning strategies using vectors containing the Z or Z peptide gene.CAS Number: 367-93-1Molecular Weight: 238.3 DaChemical Formula: C9H18O5SSolubility: Soluble in water (5%), renders clear colorless solutionStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.