Learn More
IBI SCIENTIFIC 3P IPTG | 367-93-1 | MFCD00063273 | 5 g
Supplier:  IBI SCIENTIFIC 3P IB02105
Encompass_Preferred
IPTG - 5gm IPTG is an inducer of B-galactosidase activity in E. coli. It is commonly used in conjunction with X-GAL to detect lac gene expression in cloning applications, thus allowing detection of recominant molecules.
Catalog No. NC0753450