Learn More
Enzo Life Sciences IPTG (animal free) (free of dioxane) (5g). CAS: 367-93-1
SDP
Catalog No. 502009590
Encompass_Preferred
Inducer of β-D-galactosidase. Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function. Contains no animal-derived products. Purity: ≥98% (HPLC). Solubility: Soluble in water (5%) or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9590 Supplier Enzo Life Sciences Supplier No. ALX582012G005
Inducer of β-D-galactosidase. Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function. Contains no animal-derived products. Purity: ≥98% (HPLC). Solubility: Soluble in water (5%) or methanol. Long Term Storage: -20°C.