Learn More
AdipoGen IPTG (animal-free, dioxane-free), AdipoGen Life Sciences. Chemical. CAS: 367-93-1.
SDP
Catalog No. 501175907
Encompass_Preferred
IPTG (animal-free, dioxane-free), AdipoGen Life Sciences. Chemical. CAS: 367-93-1. Formula: C9H18O5S. MW: 238.3. Molecular biology reagent. beta-Galactosidase activity inducer in several bacteria, by binding and inhibiting the lac repressor. Used in gene cloning together with X-Gal (Prod. No. AG-CC1-0003), in a technique called blue/white screening. Distinguishes between recombinant and non-recombinant plasmids carrying the beta-galactosidase gene. Plant origin, contains no animal-derived contaminants. Stimulates beta-galactosidase in cellular systems in which dioxane would disrupt normal cell function. 367-93-1
Catalog No. 50-117-5907 Supplier AdipoGen Supplier No. AGCC10002G001
IPTG (animal-free, dioxane-free), AdipoGen Life Sciences. Chemical. CAS: 367-93-1. Formula: C9H18O5S. MW: 238.3. Molecular biology reagent. beta-Galactosidase activity inducer in several bacteria, by binding and inhibiting the lac repressor. Used in gene cloning together with X-Gal (Prod. No. AG-CC1-0003), in a technique called blue/white screening. Distinguishes between recombinant and non-recombinant plasmids carrying the beta-galactosidase gene. Plant origin, contains no animal-derived contaminants. Stimulates beta-galactosidase in cellular systems in which dioxane would disrupt normal cell function. 367-93-1